missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EWSR1 Recombinant Rabbit Monoclonal Antibody (23GB1590), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 186.00 - € 446.00
Specifications
| Antigen | EWSR1 |
|---|---|
| Clone | 23GB1590 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089441
![]() |
Invitrogen™
MA553122 |
100 μL |
€ 446.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089747
|
Invitrogen™
MA553123 |
20 μL |
€ 186.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EWSR1 Recombinant Monoclonal antibody specifically detects EWSR1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| EWSR1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q01844, Q61545 | |
| 14030, 2130, 289752 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1590 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AC002059.7; bK984G1.4; Ewing sarcoma breakpoint region 1; Ewing sarcoma breakpoint region 1 protein; Ewing sarcoma homolog; Ewings sarcoma EWS-Fli1 (type 1) oncogene; Ews; EWS oncogene; EWS RNA binding protein 1; EWS RNA-binding protein 1; EWS RNA-binding protein variant 6; EWS-FLI1; Ewsh; EWSR1; RNA-binding protein EWS | |
| A synthesized peptide derived from human EWSR1 (100-162AA). | |
| Primary | |
| -20°C | |
| EWSR1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title