missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXD1 Antibody (OTI5B5), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72443F
This item is not returnable.
View return policy
Description
EXD1 Monoclonal specifically detects EXD1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| EXD1 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| EXD1 | |
| Full length recombinant protein of human EXD1 (NP_689809) produced in HEK293T cells. | |
| 0.1 mL | |
| 161829.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI5B5 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| EXDL1, exonuclease 3′-5′ domain containing 1, exonuclease 3′-5′ domain-containing protein 1, exonuclease 3′-5′ domain-like 1, Exonuclease 3′-5′ domain-like-containing protein 1, MGC33637 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction