missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EXOC7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522215
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (96%), Rat (96%), Chicken (93%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a component of the exocyst complex. The exocyst complex plays a critical role in vesicular trafficking and the secretory pathway by targeting post-Golgi vesicles to the plasma membrane. The encoded protein is required for assembly of the exocyst complex and docking of the complex to the plasma membrane. The encoded protein may also play a role in pre-mRNA splicing through interactions with pre-mRNA-processing factor 19. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene, and a pseudogene of this gene is located on the long arm of chromosome 4.
Specifications
| EXOC7 | |
| Polyclonal | |
| Unconjugated | |
| EXOC7 | |
| 2-5-3p; EX070; EXO70; Exo70p; EXOC1; Exoc7; Exocyst complex component 7; exocyst complex component Exo70; exocyst component protein 70 kDa homolog; HGNC:23214; KIAA1067; rExo70; sec70; YJL085W | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23265 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q9UPT5 | |
| EXOC7 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human EXOC7. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction