missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EXOSC1 Monoclonal Antibody (OTI1H9)
GREENER_CHOICE

Product Code. 15996594
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15996594 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15996594 Supplier Invitrogen™ Supplier No. MA527008

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

EXOSC1 Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a core component of the exosome. The mammalian exosome is required for rapid degradation of AU rich element-containing RNAs but not for poly(A) shortening. The association of this protein with the exosome is mediated by protein-protein interactions with ribosomal RNA-processing protein 42 and ribosomal RNA-processing protein 46.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXOSC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1H9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EXOSC1
Gene Accession No. Q9Y3B2
Gene Alias 2610035C18Rik; 2610104C07Rik; 2610312F07Rik; 3'-5' exoribonuclease CSL4 homolog; AI447561; CGI-108; CSL4; Csl4p; EXOSC1; exosomal core protein CSL4; exosome complex component CSL4; Exosome component 1; hCsl4p; homolog of yeast exosomal core protein CSL4; p13; RP11-452K12.9; SKI4; Ski4p
Gene Symbols EXOSC1
Host Species Mouse
Immunogen Full length human recombinant protein of EXOSC1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51013
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.