missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exosome component 1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22239UV
This item is not returnable.
View return policy
Description
Exosome component 1 Polyclonal antibody specifically detects Exosome component 1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Exosome component 1 | |
| Polyclonal | |
| Western Blot | |
| CGI-108,3'-5' exoribonuclease CSL4 homolog, CSL4 exosomal core protein homolog, Csl4p, CSL4p13, exosomal core protein CSL4, exosome component 1hCsl4p, homolog of yeast exosomal core protein CSL4, SKI4, Ski4p | |
| The immunogen this antibody was made to, maps to a region between residue 145 and 195 of human Exosome Component 1 using the numbering given in entry NP_057130.1 (GeneID 51013). | |
| 0.1 mL | |
| Immunology | |
| 51013 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction