missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exosome component 1 Antibody (OTI1H9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45545
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
Exosome component 1 Monoclonal specifically detects Exosome component 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Exosome component 1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:150 | |
| CGI-108,3'-5' exoribonuclease CSL4 homolog, CSL4 exosomal core protein homolog, Csl4p, CSL4p13, exosomal core protein CSL4, exosome component 1hCsl4p, homolog of yeast exosomal core protein CSL4, SKI4, Ski4p | |
| Mouse | |
| 21.3 kDa | |
| 0.1 mL | |
| Immunology | |
| 51013 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1H9 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| EXOSC1 | |
| Full length human recombinant protein of human EXOSC1(NP_057130) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction