missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exosome component 10 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97516AF532
This item is not returnable.
View return policy
Description
Exosome component 10 Polyclonal specifically detects Exosome component 10 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Exosome component 10 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| autoantigen PM-SCL, EC 3.1.13.-, exosome component 10, P100 polymyositis-scleroderma overlap syndrome-associated autoantigen, p2, p3, p4, PM/Scl-100Autoantigen PM/Scl 2, PM-Scl, PMSCL2PMSCL, Polymyositis/scleroderma autoantigen 2, polymyositis/scleroderma autoantigen 2 (100kD), polymyositis/scleroderma autoantigen 2, 100kDa, RRP6, Rrp6p, RRP6Polymyositis/scleroderma autoantigen 100 kDa | |
| Produced in rabbits immunized with E. coli-derived Human Exosome component 10 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 5394 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction