missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Exosome component 4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30528
This item is not returnable.
View return policy
Description
Exosome component 4 Polyclonal antibody specifically detects Exosome component 4 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Exosome component 4 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| exosome complex exonuclease RRP41, exosome component 4Rrp41p, exosome component Rrp41, FLJ20591, p12ARibosomal RNA-processing protein 41, RRP41A, RRP41Ski6p, SKI6hRrp41p | |
| Between 1 and 50 | |
| 100 μg | |
| DNA replication Transcription Translation and Splicing | |
| 54512 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction