missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exosome component 5 Antibody (V80P1C10*C), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50173AF488
This item is not returnable.
View return policy
Description
Exosome component 5 Monoclonal specifically detects Exosome component 5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.
Specifications
| Exosome component 5 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50 mM Sodium Borate with 0.05% Sodium Azide | |
| EXOSC5 | |
| Peptide Sequence - KLLMSSTKGLY | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing, Epigenetics | |
| 56915 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| V80P1C10*C | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| CML28, exosome complex exonuclease RRP46, exosome component 5Chronic myelogenous leukemia tumor antigen 28, exosome component Rrp46, hRrp46p, MGC12901, p12BMGC111224, RRP41B, Rrp46p, RRP46Ribosomal RNA-processing protein 46 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction