missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exosome component 7 Antibody (SR1527), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22416-50ul
This item is not returnable.
View return policy
Description
Exosome component 7 Monoclonal antibody specifically detects Exosome component 7 in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
| Exosome component 7 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| SR1527 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| EAP1, exosome complex exonuclease RRP42, exosome component 7Rrp42p, KIAA0116hRrp42p, p8FLJ26543, RRP42Ribosomal RNA-processing protein 42 | |
| A synthesized peptide derived from Exosome component 7 (Uniprot #: Q15024) | |
| 50 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 23016 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction