missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exosome component 8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-47465-25ul
This item is not returnable.
View return policy
Description
Exosome component 8 Polyclonal specifically detects Exosome component 8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications
| Exosome component 8 | |
| Polyclonal | |
| Western Blot 0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| bA421P11.3, CBP-interacting protein 3, CIP3, EAP2, exosome complex exonuclease RRP43, exosome component 8Opa-interacting protein 2, OIP2Ribosomal RNA-processing protein 43, Opa interacting protein 2, p9RP11-421P11.3, RRP43OIP-2, Rrp43p | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EXOSC8 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ALAEVNLKKKSYLNIRTHPVATSFAVFDDTLLIVDPTGEEEHLATGTLTIVMDEEGKLCCLHKPGGSGLTGAKLQDCMSRAVTRHKEVKKLMDEVIKSMK | |
| 25 μL | |
| DNA replication Transcription Translation and Splicing | |
| 11340 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction