missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exosome Component 9 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71702IR
This item is not returnable.
View return policy
Description
Exosome Component 9 Polyclonal antibody specifically detects Exosome Component 9 in Human, Mouse, Chicken samples. It is validated for Western Blot, Simple Western, ChIP assay, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP assay (ChIP)
Specifications
| Exosome Component 9 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 5393 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Simple Western, ChIP Assay, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DyLight 755 | |
| Autoantigen PM/Scl 1, EC 3.1.13, exosome component 9Polymyositis/scleroderma autoantigen 75 kDa, p6, PM/Scl-75P75 polymyositis-scleroderma overlap syndrome-associated autoantigen, PMSCL175kD, Polymyositis/scleroderma autoantigen 1, polymyositis/scleroderma autoantigen 1 (75kD), polymyositis/scleroderma autoantigen 1, 75kDa | |
| A genomic peptide made to an internal region of human Exosome Component 9 (within residues 250-439). [Swiss-Prot Q06265] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction