Learn More
Gibco™ Expi293F™ Inducible GnTI- Cells
Description
Cell line established by the selective knockout of the GnTI gene and stable integration of the regulatory plasmid pcDNA6/TR, which encodes the TetR gene under the control of the human CMV promoter. It is a powerful tool for tetracycline-inducible high-yield expression of homogeneously glycosylated recombinant proteins.
Features:
- Suspension, high-density culture in Expi293 Expression Medium
- Homogeneous N-glycosylation of expressed proteins
- Tetracycline-inducible protein expression with extremely low basal expression levels
- Equivalent protein yields to parental Expi293F cells—up to 1 g/L
For inducible expression of aprotein of interest, pcDNA5/TO mammalian expresson vector or an equivalent inducible vector is required for tetracycline-regulated gene expression.
Frozen cells are supplied in a vial containing 1 mL of cells at 1 x 107 viable cells/mL in Expi293 Expression Medium and 10% DMSO. The cells should be thawed directly into Expi293 Expression Medium.
Specifications
Specifications
| Content And Storage | 1 vial Expi293F Inducible GnTI- Cells (1x107 cells/vial), store in vapor-phase liquid nitrogen |
| Product Line | Expi293 |
| Quantity | 1 vial |
| Shipping Condition | Dry Ice |