missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exportin-5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-89606-25ul
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Exportin-5 Polyclonal specifically detects Exportin-5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Especificaciones
| Exportin-5 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:200 - 1:500, Knockdown Validated | |
| Q9HAV4 | |
| XPO5 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ILGPLFTYLHMRLSQKWQVINQRSLLCGEDEAADENPESQEMLEEQLVRMLTREVMDLITVCCVSKKGADHSSAPPADGDDEEMMATEVTPSAMAELTDLGKCLMKHEDVCTALLITAFNSLAWKDTLSC | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of human Exportin-5 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Exp5, exportin 5, exportin-5, FLJ14239, FLJ32057, FLJ45606, KIAA1291exp5, Ran-binding protein 21, RANBP21 | |
| Rabbit | |
| 136 kDa | |
| 25 μL | |
| Epigenetics, microRNA | |
| 57510 | |
| Human, Mouse, Rat | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido