missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ext2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35904-20ul
This item is not returnable.
View return policy
Description
Ext2 Polyclonal antibody specifically detects Ext2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| Ext2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| EC 2.4.1.224, EC 2.4.1.225, exostoses (multiple) 2, exostosin 2, exostosin-2, Glucuronosyl-N-acetylglucosaminyl-proteoglycan/N-acetylglucosaminyl-proteoglycan4-alpha-N-acetylglucosaminyltransferase, Multiple exostoses protein 2, N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase, Putative tumor suppressor protein EXT2, SOTV | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 80-340 of human Ext2 (NP_000392.3).,, Sequence:, PHSIESSNDWNVEKRSIRDVPVVRLPADSPIPERGDLSCRMHTCFDVYRCGFNPKNKIKVYIYALKKYVDDFGVSVSNTISREYNELLMAISDSDYYTDDINRACLFVPSIDVLNQNTLRIKETAQAMAQLSRWDRGTNHLLFNMLPGGPPDYNTALDVPRDRALLAGGGFSTWTYRQGYDVSIPVYSPLSAEVDLPEKGPGPRQYFLLSSQVGLHPEYREDLEALQVKHGESVLVLDKCTNLSEGVLSVRKRCHKHQVFD | |
| 20 μL | |
| Cell Cycle and Replication | |
| 2132 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction