Learn More
Memmert™ Extended Overtemperature Protection for IF450
Product Code. 17516975 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IF450 Incubator
Unit Size:
Each
Product Code. 17516975
Brand: Memmert™ IF450A6
Only null left
Add to basket
Add to basket
Extended overtemperature protection with integrated Pt100 sensor for independent temperature monitoring.
  • Accessory
  • With integrated Pt100 sensor
  • For models with SingleDISPLAY
Specifications
For Use With (Equipment) IF450 Incubator
Includes Integrated Pt100 Sensor