Learn More
Memmert™ Extended Overtemperature Protection for IF55
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IF55 Incubator
Unit Size:
Each
Description
- Accessory
- With integrated Pt100 sensor
- For models with SingleDISPLAY
Specifications
Specifications
| For Use With (Equipment) | IF55 Incubator |
| Includes | Integrated Pt100 Sensor |