Learn More
Memmert™ Extended Overtemperature Protection for SF75
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
SF75 Sterilizer
Unit Size:
Each
Description
- Accessory
- With integrated Pt100 sensor
- For models with SingleDISPLAY
Specifications
Specifications
| For Use With (Equipment) | SF75 Sterilizer |
| Includes | Integrated Pt100 Sensor |