Learn More
Ismatec™ Extension Tubing, Tygon ST
missing translation for 'orderingAttributeHoverText'
Tubing Size:
I.D. 0.19 mm
I.D. 0.38 mm
I.D. 0.57 mm
I.D. 0.89 mm
I.D. 1.14 mm
I.D. 1.65 mm
I.D. 1.85 mm
missing translation for 'unitSize'
10m
Each
Description
- Inexpensive, all-around tubing for general laboratory applications.
- Transparent
- Resistant to almost all inorganic chemicals
- Tasteless
- Smooth, polished inner wall
- Low gas permeability
- Non-aging and non-oxidizing
- High dialectric constant
Specifications
Specifications
| Certifications/Compliance | FDA 21 CFR175.300 |
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 1.65 mm |
| Material | Tygon ST R-3603, R-3607 |
| Overall Length (Metric) | 10 m |
| For Use With (Application) | General laboratory applications, 2- and 3-stop color-coded tubing. |
| Min. Operating Temperature (Metric) | −50°C |
| Packaging Quantity | 1 Reel |
| Resistance Properties | Acids: Good, Alkaline solutions: Good, Solvents: Not rec., Pressure: Fair, Vacuum: Good, Viscous media: Excellent, Sterile media: Limited |
| Tubing Size | I.D. 1.65 mm |
| Show More |