missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EXTL2 Monoclonal Antibody (339509)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA561170

Product Code. 30535524

  • € 429.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

EXTL2 is a glycosyltransferase required for the biosynthesis of heparan-sulfate and responsible for the alternating addition of beta-1-4-linked glucuronic acid (GlcA) and alpha-1-4-linked N-acetylglucosamine (GlcNAc) units to nascent heparan sulfate chains.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EXTL2
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q9ES89
EXTL2
Mouse myeloma cell line NS0-derived recombinant mouse Exostosin-like 2/EXTL2, Asn43-Met330.
100 μg
Primary
Mouse
Antibody
IgG2a
Western Blot
339509
Unconjugated
EXTL2
3000001D04Rik; alpha-1,4-N-acetylhexosaminyltransferase EXTL2; Alpha-GalNAcT EXTL2; AW146439; exostoses (multiple)-like 2; exostoses-like 2; exostosin like glycosyltransferase 2; exostosin-like 2; exostosin-like glycosyltransferase 2; exotoses (multiple)-like 2; EXTL2; Extr2; EXT-related protein 2; Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; processed exostosin-like 2
Rat
Protein A/G
RUO
58193
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.