missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EXTL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550406
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total EXTL3 protein.
EXTL3 is probable glycosyltransferase.
Specifications
| EXTL3 | |
| Polyclonal | |
| Unconjugated | |
| EXTL3 | |
| 2900009G18Rik; AU040105; BOTV; exostoses (multiple)-like 3; exostoses-like 3; exostosin; exostosin like glycosyltransferase 3; exostosin tumor-like 3; exostosin-like 3; exostosin-like glycosyltransferase 3; Ext1l; EXTL1L; EXTL3; EXTR1; EXT-related 1; EXT-related protein 1; Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; hereditary multiple exostoses gene isolog; KIAA0519; mKIAA0519; multiple exostosis-like protein 3; Putative tumor suppressor protein EXTL3; REG; reg receptor; REGR; RPR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2137, 54616 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.2 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O43909, Q9WVL6 | |
| EXTL3 | |
| Fusion protein corresponding to residues near the C terminal of human exostosin-like glycosyltransferase 3. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction