Learn More
Lenz Laborglasintrumente Extractors Acc. to Twisselmann
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05380031
Additional Details : Weight : 0.50000kg
Description
- Extractor head with vacuum jacket
- Made of DURAN™ tubing
- Twisselmann condenser (improved) with thread GL 14
- Round bottom flask
Specifica
70 mL | |
Duran Glass | |
NS 34/35 |