Learn More
Lenz Laborglasintrumente Extractors for Specific Light Solvents
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marke: Lenz Laborglasintrumente 05432037
Weitere Details : Weight : 10.00000kg
Beschreibung
- Extractor head with insert filter (frit, porosity: 0)
- With PTFE valve stopcock
- Made of DURAN™ tubing
- Dimroth condenser
- With thread GL 14
- Round bottom flask
Spezifikation
250 mL | |
0 | |
NS 45/40 |