Learn More
Karl Hecht Extraction attachments 250ml
Brand:  Karl Hecht 41155036
Extraction attachments 250ml
Product Code. 10195471
  • ONLINE EXCLUSIVE PRICE
    € 136.50
    € 91.84 / Each
    Save € 44.66
    32% Off
Only null left
Estimated Shipment: 19-02-2026