missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EYA2 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-46816APC
This item is not returnable.
View return policy
Description
EYA2 Polyclonal antibody specifically detects EYA2 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot (Negative)
Specifications
| EYA2 | |
| Polyclonal | |
| Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative) | |
| EAB1EC 3.1.3.48, eyes absent (Drosophila) homolog 2, eyes absent homolog 2, eyes absent homolog 2 (Drosophila), MGC10614 | |
| The immunogen for this product maps to a region between residue 1 and 50 of human eyes absent homolog 2 using the numbering given in entry NP_005235.3 (GeneID 2139). | |
| 0.1 mL | |
| Protein Phosphatase, Vision | |
| 2139 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction