Learn More
Plum™ Eye Emergency Station
Product Code. 10445792
Change view
Click to view available options
Capacity (Metric):
200 mL, 500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10445792 200 mL, 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10445792 Supplier Plum™ Supplier No. 4770
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
With 200 mL pH Neutral Eyewash and 500 mL Eye Rinse
  • Includes 1 saline solution 0.9% (500 mL) and 1 phosphate solution 4.9% (200 mL)
  • For rinsing foreign objects (saline solution) and treatment of acid and alkali exposure (phosphate solution)
  • Flushing Duration: 5min (saline solution), 2min (phosphate solution)
  • Features eye support for one eye, with wall mount
Specifications
Capacity (Metric) 200 mL, 500 mL
Includes 200 mL pH Neutral Eyewash and 500 mL Eye Rinse