Learn More
Ekastu Safety™ EPS Eye Wash Station Kit
Supplied complete with 2 EKASTU Eye Wash Bottles. EKASTU SAfety™ EPS Eye Wash Station Kit is dustproof with a pull-top lid and protects Eye Wash Bottles from over exposure to warm and cold temperatures.
Brand:  Ekastu Safety™ 277999
Additional Details : Weight : 0.10000kg
Product Code. 10789175
  • € 71.95 / Each
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
  • For direct availability in danger areas
  • Dust-proof with a pull-top lid
  • Protects Eye Wash Bottles from cold and warmth
  • Date of expiry can be stated on the lid
  • Bottles can be stored -maintenance free- up to three years
  • Easy installation
  • Consisting of: EKASTU Eye Wash Bottle ECO with funnel, 600mL FD; and EKASTU Eye Wash Bottle MINI-ECO with funnel, 200mL FD
Specifications
Specifications
Storage of Eye Wash Bottles
EKASTU Eye Wash Bottle ECO with Funnel, 600 ml FD; and EKASTU Eye Wash Bottle MINI-ECO with funnel, 200 ml FD
Wall
Eye Wash Station