missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EZH2/KMT6 Antibody (CL11915) [Alexa Fluor™ 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21204AF594
This item is not returnable.
View return policy
Description
EZH2/KMT6 Monoclonal antibody specifically detects EZH2/KMT6 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| EZH2/KMT6 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| EC 2.1.1, EC 2.1.1.43, enhancer of zeste (Drosophila) homolog 2, enhancer of zeste 2, Enhancer of zeste homolog 2, enhancer of zeste homolog 2 (Drosophila), ENX1, ENX-1KMT6EZH1KMT6A, histone-lysine N-methyltransferase EZH2, Lysine N-methyltransferase 6, MGC9169 | |
| This antibody was developed using a synthetic peptide derived from Q15910, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Chromatin Research, Phospho Specific, Prostate Cancer | |
| 2146 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL11915 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction