Learn More
EZH2/KMT6 Antibody (EZH2/2536), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-08932AF532
Code produit. 18504196
  • € 595.00 / 100µL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
EZH2/KMT6 Monoclonal antibody specifically detects EZH2/KMT6 in Human samples. It is validated for Western Blot, Protein Array
TRUSTED_SUSTAINABILITY
Spécification
Spécification
EZH2/KMT6
Monoclonal
Alexa Fluor 532
EC 2.1.1, EC 2.1.1.43, enhancer of zeste (Drosophila) homolog 2, enhancer of zeste 2, Enhancer of zeste homolog 2, enhancer of zeste homolog 2 (Drosophila), ENX1, ENX-1KMT6EZH1KMT6A, histone-lysine N-methyltransferase EZH2, Lysine N-methyltransferase 6, MGC9169
Recombinant full-length human EZH2/KMT6 protein (Uniprot: Q15910)
100 μg
Breast Cancer, Cancer, Chromatin Research, Prostate Cancer
2146
Store at 4°C in the dark.
IgG2a κ
Western Blot, Protein Array
EZH2/2536