missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EZH2/KMT6 Antibody (EZH2/6988) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20982
This item is not returnable.
View return policy
Description
EZH2/KMT6 Monoclonal antibody specifically detects EZH2/KMT6 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry (Paraffin)
Specifications
| EZH2/KMT6 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| EC 2.1.1, EC 2.1.1.43, enhancer of zeste (Drosophila) homolog 2, enhancer of zeste 2, Enhancer of zeste homolog 2, enhancer of zeste homolog 2 (Drosophila), ENX1, ENX-1KMT6EZH1KMT6A, histone-lysine N-methyltransferase EZH2, Lysine N-methyltransferase 6, MGC9169 | |
| Recombinant fragment (around aa300-550) of human EZH2/KMT6 protein (exact sequence is proprietary) | |
| 100 μg | |
| Breast Cancer, Cancer, Chromatin Research, Phospho Specific, Prostate Cancer | |
| 2146 | |
| Store at -20°C to -70°C. Avoid freeze/thaw cycles. | |
| IgG κ |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry (Paraffin) | |
| EZH2/6988 | |
| Unconjugated | |
| 10 mM PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction