missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ezrin Antibody (2086A) [PE], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB7239P
This item is not returnable.
View return policy
Description
Ezrin Monoclonal antibody specifically detects Ezrin in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry
Specifications
| Ezrin | |
| Monoclonal | |
| PE | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry | |
| 2086A | |
| Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
| CVIL, CVL, Cytovillin, cytovillin 2, DKFZp762H157, EZR, ezrin, FLJ26216, p81, VIL2MGC1584, villin 2 (ezrin), villin-2 | |
| E. coli-derived recombinant human Ezrin, Lys438-Arg562, Accession # P15311 | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Signal Transduction | |
| 7430 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction