Learn More
Ezrin Antibody (2086C), Alexa Fluor™ 750, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals FAB72391S
Product Code. 30063140
  • € 618.00 / 0.10mL
Description
Ezrin Monoclonal antibody specifically detects Ezrin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ezrin
Monoclonal
Alexa Fluor 750
50mM Sodium Borate
Rabbit
Protein A or G purified from cell culture supernatant
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry
2086C
Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry
CVIL, CVL, Cytovillin, cytovillin 2, DKFZp762H157, EZR, ezrin, FLJ26216, p81, VIL2MGC1584, villin 2 (ezrin), villin-2
E. coli-derived recombinant human Ezrin, Lys438-Arg562, Accession # P15311
0.1 mL
Cancer, Cell Cycle and Replication, Phospho Specific, Signal Transduction
7430
Store at 4C in the dark.