Learn More
R&DSystems™ F-12, 25 mM HEPES, L-glutamine
Product Code. 18461210
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. 18461210
Brand: R&DSystems™ M25250
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ham’s Nutrient Mixture F-12 with 25 mM HEPES buffer and L-glutamine.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Cell Culture
Quantity 500 mL
Purity or Quality Grade 0.2 μm filtered
Content And Storage 2 - 8 °C, protected from light