Learn More
F(ab')2-Rabbit anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, Invitrogen™
F(ab')2-Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen 31109
Additional Details : Weight : 0.80000kg
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31109 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31109 reacts with the heavy chains of goat IgG and with the light chains common to most goat immunoglobulins, but does not react against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with human serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of origin: USA NOTE: BSA and dry milk may contain IgG which reacts with anti-bovine IgG, anti-goat IgG and anti-sheep IgG antibodies. Use of BSA and/or dry milk may significantly increase background and/or reduce secondary antibody titer.
Goat IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Goat IgG (H+L) Cross-Adsorbed in Goat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Specifications
Goat IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Unconjugated | |
Rabbit | |
500 μg | |
Secondary | |
4° C | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.6 mg/mL | |
PBS with no preservative; pH 7.6 | |
Antigen affinity chromatography | |
RUO | |
Goat | |
Liquid |