missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F12 Monoclonal Antibody (OTI1H5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807219S
This item is not returnable.
View return policy
Description
F12 Monoclonal antibody specifically detects F12 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| F12 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q12888 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7158 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| OTI1H5 | |
| Unconjugated | |
| TP53BP1 | |
| beta-factor XIIa part 1; Beta-factor XIIa part 2; coagulation factor XII; coagulation factor XII (Hageman factor); coagulation factor XIIa heavy chain; Coagulation factor XIIa light chain; F12; F13; Factor 12; factor XII; Factor12; FXII; HAE3; HAEX; HAF; hageman factor | |
| Human recombinant protein fragment corresponding to amino acids 1-300 of human TP53BP1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction