missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F2 Monoclonal Antibody (OTI5B11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812512
This item is not returnable.
View return policy
Description
Coagulation factor II is proteolytically cleaved to form thrombin in the first step of the coagulation cascade which ultimately results in the stemming of blood loss. F2 also plays a role in maintaining vascular integrity during development and postnatal life. Mutations in F2 leads to various forms of thrombosis and dysprothrombinemia.Specifications
| F2 | |
| Monoclonal | |
| Unconjugated | |
| F2 | |
| PT, RPRGL2, THPH1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2147 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI5B11 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P00734 | |
| F2 | |
| Human recombit protein fragment corresponding to amino acids 44-622 of human F2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction