Learn More
F2 Mouse anti-Human, Clone: OTI5G10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF812513
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Coagulation factor II is proteolytically cleaved to form thrombin in the first step of the coagulation cascade which ultimately results in the stemming of blood loss. F2 also plays a role in maintaining vascular integrity during development and postnatal life. Mutations in F2 leads to various forms of thrombosis and dysprothrombinemia.Specifications
| F2 | |
| Monoclonal | |
| Unconjugated | |
| F2 | |
| PT, RPRGL2, THPH1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2147 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI5G10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P00734 | |
| F2 | |
| Human recombit protein fragment corresponding to amino acids 44-622 of human F2 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.