Learn More
F4/80 Antibody (521204) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB5580JF635
Product Code. 30171967
  • € 617.00 / 0.10mL
Description
F4/80 Monoclonal antibody specifically detects F4/80 in Mouse samples. It is validated for Flow Cytometry,Immunocytochemistry,CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
F4/80
Monoclonal
Janelia Fluor 635
ADGRE1, cell surface glycoprotein F4/80, DD7A5-, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, Emr, Emr1, EMR1 hormone receptor, F4/80, Gpf480, Ly7, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7L, TM7LN3
NS0 mouse myeloma cell line transfected with mouse F4/80/EMR1, Gln28-Gly931, Accession # Q61549
RUO
Primary
Mouse
Liquid
Flow Cytometry, Immunocytochemistry, CyTOF
521204
50mM Sodium Borate
Rat
0.1 mL
Adaptive Immunity, Cellular Markers, Immune System Diseases, Immunology, Innate Immunity
13733
Store at 4°C in the dark.
IgG2a