missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F4/80 Antibody (Cl:A3-1 (recombinant version)), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81029DL594
This item is not returnable.
View return policy
Description
F4/80 Monoclonal specifically detects F4/80 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
| F4/80 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Adgre1 | |
| Thioglycollate stimulated peritoneal macrophages of mouse origin. | |
| 0.1 mL | |
| Adaptive Immunity, GPCR, Immune System Diseases, Immunology, Innate Immunity, Myeloid derived Suppressor Cell | |
| 13733 | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry | |
| Cl:A3-1 (recombinant version) | |
| Flow Cytometry, Immunohistochemistry | |
| ADGRE1, Adhesion G protein-coupled receptor E1, cell surface glycoprotein F4/80, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, EMR1 hormone receptor, F4/80, Gpf480, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7LN3 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody recognises the mouse F4/80 antigen, a 160kD glycoprotein, expressed by murine macrophages. | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction