Learn More
F4/80 Antigen Rat, PE-Cy7, Clone: BM8.1, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB68-PE7-100ug
 View more versions of this product
Product Code. 15902368
  • € 592.20 / 100µg
Specifications
F4/80 Antigen
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Adgre1
Protein A/G Purified
RUO
13733
IgG2b κ
Flow Cytometry
BM8.1
PE-Cyanine7
XP_006523664
Rat
100 μg
Primary
Store at 2-4°C.