Learn More
F4/80 Rat, anti-Mouse, Unconjugated, Clone: BM8, STEMCELL Technologies
Rat Monoclonal Antibody
Brand:  Stemcell Technologies 60027
Encompass_Preferred
Product Code. 17178581
  • € 537.30 / 500µg
Description
  • The BM8 antibody reacts with the F4/80 antigen (also termed Ly-71 in mouse and EMR-1 in human), an ∼160 kDa transmembrane glycoprotein belonging to the EGF-TM7 family of G-protein-coupled receptors
  • F4/80 is considered a marker of choice for the identification of mature tissue macrophages, being broadly but variably expressed by this cell type in the liver (Kupffer cells), skin (Langerhans cells), bone marrow stroma, pancreas, thymus, spleen (red pulp), lung, and other tissues
  • It is also expressed by circulating monocytes, eosinophils and a subset of dendritic cells
  • F4/80 is reportedly the only macrophage marker suitable for distinguishing destructive from non-destructive inflammatory processes in the pancreas
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Monocyte Enrichment Kit (Catalog #19761) and EasySep™ Mouse CD11b Positive Selection Kit (Catalog #18770)
Specifications
Specifications
F4/80
Monoclonal
Unconjugated
500 μg
Human, Mouse
Monocytes, Other
Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry
BM8
Rat
Primary
IgG2a κ