Learn More
Invitrogen™ F4/80 Recombinant Rabbit Monoclonal Antibody (4J7L6)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555206
Description
Immunogen Sequence: KRILFKCKED LILQSEQIQQ CQAVQGRDLG YASFCTLVNA TFTILDNTCE NKSAPVSLQS AATSVSLVLE QATTWFELSK EETSTLGTIL LETVESTMLA ALLIPSGNAS QMIQTEYLDI ESKVINEECK ENESINLAAR GDKMNVGCFI IKESVSTGAP GVAFVSFAHM ESVLNERFFE DGQSFRKLRM NSRVVGGTVT GEKKEDFSKP IIYTLQHIQP KQKSERPICV SWNTDVEDGR WTPSGCEIVE ASETHTVCSC NRMANLAIIM ASGELTMEF.
F4/80 antigen is a 160 kDa cell surface glycoprotein that is a member of the EGF TM7 family of proteins and shares 68% overall amino acid identity with human EGF module containing mucin like hormone receptor 1 (EMR1). Expression of F4/80 is heterogeneous and is reported to vary during macrophage maturation and activation. The F4/80 antigen is expressed on a wide range of mature tissue macrophages including Kupffer cells, Langerhans, microglia, macrophages located in the gut lamina propria, peritoneal cavity, lung, thymus, bone marrow stroma and macrophages in the red pulp of the spleen. F4/80 expression has also been reported on a subpopulation of dendritic cells but is absent from macrophages located in T cell areas of the spleen and lymph node. The ligands and biological functions of the F4/80 antigen have not yet been determined but recent studies suggest a role for F4/80 in the generation of efferent CD8+ve regulatory T cells.
Specifications
| F4/80 | |
| Recombinant Monoclonal | |
| 1.6 mg/mL | |
| PBS with 50% glycerol, 0.05% BSA and 0.05% ProClin 300; pH 7.3 | |
| Q61549 | |
| ADGRE1 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 4J7L6 | |
| Unconjugated | |
| ADGRE1 | |
| ADGRE1; Adhesion G protein-coupled receptor E1; Adhesion G protein-coupled receptor E2; cell surface glycoprotein EMR1; cell surface glycoprotein F4/80; DD7A5-7; EGF-like module containing, mucin-like, hormone receptor-like 1; EGF-like module containing, mucin-like, hormone receptor-like sequence 1; EGF-like module receptor 1; EGF-like module-containing mucin-like hormone receptor-like 1; EGF-TM7; Emr1; EMR1 hormone receptor; F4/80; Gpf480; Ly71; lymphocyte antigen 71; TM7LN3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13733 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.