Learn More
F7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 23058-1-AP-150UL
 View more versions of this product
Product Code. 16852175
  • € 410.00 / 150µL
Description
Coagulation factor VII is a vitamin K-dependent factor essential for hemostasis. This factor circulates in the blood in a zymogen form, and is converted to an active form by either factor IXa, factor Xa, factor XIIa, or thrombin by minor proteolysis. Upon activation of the factor VII, a heavy chain containing a catalytic domain and a light chain containing 2 EGF-like domains are generated, and two chains are held together by a disulfide bond. In the presence of factor III and calcium ions, the activated factor then further activates the coagulation cascade by converting factor IX to factor IXa and/or factor X to factor Xa. Defects in F7 production can cause coagulopathy.
Specifications
Specifications
F7
Polyclonal
Unconjugated
F7
Coagulation factor VII, Eptacog alfa, F7, Proconvertin, SPCA
Rabbit
Antigen Affinity Chromatography
RUO
14068, 2155
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P08709, P70375
F7
F7 Fusion Protein Ag19324
150 μL
Primary
Human, Mouse