missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23058-1-AP-20UL
This item is not returnable.
View return policy
Description
Coagulation factor VII is a vitamin K-dependent factor essential for hemostasis. This factor circulates in the blood in a zymogen form, and is converted to an active form by either factor IXa, factor Xa, factor XIIa, or thrombin by minor proteolysis. Upon activation of the factor VII, a heavy chain containing a catalytic domain and a light chain containing 2 EGF-like domains are generated, and two chains are held together by a disulfide bond. In the presence of factor III and calcium ions, the activated factor then further activates the coagulation cascade by converting factor IX to factor IXa and/or factor X to factor Xa. Defects in F7 production can cause coagulopathy.Specifications
| F7 | |
| Polyclonal | |
| Unconjugated | |
| F7 | |
| Coagulation factor VII, Eptacog alfa, F7, Proconvertin, SPCA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14068, 2155 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P08709, P70375 | |
| F7 | |
| F7 Fusion Protein Ag19324 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction