missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FA2H Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00079152-B01P
This item is not returnable.
View return policy
Description
FA2H Polyclonal antibody specifically detects FA2H in Human samples. It is validated for Western Blot
Specifications
| FA2H | |
| Polyclonal | |
| PBS (pH 7.4) | |
| FAAHFAXDC1, FAH1, fatty acid 2-hydroxylase, Fatty acid alpha-hydroxylase, fatty acid hydroxylase domain containing 1, FLJ25287, SCS7, spastic paraplegia 35 (autosomal recessive), SPG35 | |
| FA2H (AAH17049.2, 1 a.a. - 372 a.a.) full-length human protein. MAPAPPPAASFSPSEVQRRLAAGACWVRRGARLYDLSSFVRHHPGGEQLLRARAGQDISADLDGPPHRHSANARRWLEQYYVGELRGEQQGSMENEPVALEETQKTDPAMEPRFKVVDWDKDLVDWRKPLLWQVGHLGEKYDEWVHQPVTRPIRLFHSDLIEGLSKTVWYSVPIIWVPLVLYLSWSYYRTFAQGNVRLFTSFTTEYTVAVPKSMFPGLFMLGTFLWSLIEYLIHRFLFHMKPPSDSYYLIMLHFVMHGQHHKAPFDGSRLVFPPVPASLVIGVFYLCMQLILPEAVGGTVFAGGLLGYVLYDMTHYYLHFGSPHKGSYLYSLKAHHVKHHFAHQKSGFGISTKLWDYCFHTLTPEKPHLKTQ | |
| 0.05 mg | |
| Lipid and Metabolism | |
| 79152 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| AAH17049.2 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction