missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FA2H Antibody (OTI2E3), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72192AF700
This item is not returnable.
View return policy
Description
FA2H Monoclonal specifically detects FA2H in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| FA2H | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FA2H | |
| Full length recombinant protein of human FA2H (NP_077282) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI2E3 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| FAAHFAXDC1, FAH1, fatty acid 2-hydroxylase, Fatty acid alpha-hydroxylase, fatty acid hydroxylase domain containing 1, FLJ25287, SCS7, spastic paraplegia 35 (autosomal recessive), SPG35 | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 79152.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction