missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAAH2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106978
This item is not returnable.
View return policy
Description
FAAH2 Polyclonal Antibody for Western Blot
FAAH2 encodes a fatty acid amide hydrolase that shares a conserved protein motif with the amidase signature family of enzymes. The encoded enzyme is able to catalyze the hydrolysis of a broad range of bioactive lipids, including those from the three main classes of fatty acid amides; N-acylethanolamines, fatty acid primary amides and N-acyl amino acids. This enzyme has a preference for monounsaturated acyl chains as a substrate.
Specifications
| FAAH2 | |
| Polyclonal | |
| Unconjugated | |
| FAAH2 | |
| AMDD; amidase domain containing; amidase domain-containing protein; Anandamide amidohydrolase 2; FAAH2; fatty acid amide hydrolase 2; fatty-acid amide hydrolase 2; Oleamide hydrolase 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 158584 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q6GMR7 | |
| FAAH2 | |
| Synthetic peptide directed towards the C-terminal region of human FAAH2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction