missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F(ab')2-Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Pacific Orange™, Invitrogen™
F(ab')2-Goat Polyclonal Secondary Antibody
Brand: Invitrogen P31585
Additional Details : Weight : 0.00450kg
Description
Mouse IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG (H+L) Cross-Adsorbed in Mouse samples. It is validated for Flow Cytometry.Specifications
Mouse IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Pacific Orange | |
Goat | |
purified | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
2 mg/mL | |
PBS with 5mM sodium azide; pH 7.5 | |
Gamma Immunoglobins Heavy and Light chains | |
250 μL | |
Secondary | |
4° C, store in dark | |
IgG |