missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP1/L-FABP Antibody (FABP1/9085R), Alexa Fluor™ 750, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24046AF750
This item is not returnable.
View return policy
Description
FABP1/L-FABP Monoclonal antibody specifically detects FABP1/L-FABP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FABP1/L-FABP | |
| Monoclonal | |
| Alexa Fluor 750 | |
| FABPL, fatty acid binding protein 1, liver, fatty acid-binding protein, liver, L-FABPFatty acid-binding protein 1, Liver-type fatty acid-binding protein | |
| Human recombinant FABP1/L-FABP protein fragment (around aa1-127) (exact sequence is proprietary) | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism | |
| 2168 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| FABP1/9085R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction