Learn More
Invitrogen™ FABP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ 720242
Description
These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Monkey, Cat and Pig.
FABP1 encodes the fatty acid binding protein found in liver. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABP1 and FABP6 (the ileal fatty acid binding protein) are also able to bind bile acids. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism.
Specifications
| FABP1 | |
| Polyclonal | |
| Unconjugated | |
| FABP1 | |
| 14 kDa selenium-binding protein; FABP1; FABPL; Fabplg; fatty acid binding protein 1; fatty acid binding protein 1, liver; fatty acid binding protein liver; fatty acid-binding protein 1; Fatty acid-binding protein, liver; L-FABP; Liver-type fatty acid-binding protein; p14; SCP; SCP.; Squalene- and sterol-carrier protein; Z-protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14080, 2168, 24360 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2 | |
| P02692, P07148, P12710 | |
| FABP1 | |
| Peptides corresponding to Human FABP1 (aa 70-80, 27-40). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.